Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607189 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS8 - regulator of G-protein signaling 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102450.2 | 3132 | UTR 3 | NP_001095920.1 | |||
NM_033345.3 | 3132 | UTR 3 | NP_203131.1 | |||
XM_005245555.3 | 3132 | Intron | XP_005245612.1 | |||
XM_011510089.2 | 3132 | UTR 3 | XP_011508391.1 | |||
XM_011510090.2 | 3132 | UTR 3 | XP_011508392.1 | |||
XM_011510091.2 | 3132 | Intron | XP_011508393.1 | |||
XM_017002631.1 | 3132 | UTR 3 | XP_016858120.1 | |||
XM_017002632.1 | 3132 | UTR 3 | XP_016858121.1 | |||
XM_017002633.1 | 3132 | UTR 3 | XP_016858122.1 | |||
XM_017002634.1 | 3132 | UTR 3 | XP_016858123.1 | |||
XM_017002635.1 | 3132 | UTR 3 | XP_016858124.1 | |||
XM_017002636.1 | 3132 | Intron | XP_016858125.1 | |||
XM_017002637.1 | 3132 | Intron | XP_016858126.1 |
Set Membership: |
JSNP |