Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAGCTAAAGAGACTTTTGCAAT[A/G]TATTTTTTCAGAGTCAGCCTTGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD45 - ankyrin repeat domain 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198493.2 | 892 | Silent Mutation | TAC,TAT | Y,Y 174 | NP_940895.1 | |
XM_011509466.2 | 892 | Silent Mutation | TAC,TAT | Y,Y 174 | XP_011507768.1 | |
XM_011509467.1 | 892 | Silent Mutation | TAC,TAT | Y,Y 174 | XP_011507769.1 | |
XM_011509468.1 | 892 | Silent Mutation | TAC,TAT | Y,Y 174 | XP_011507770.1 | |
XM_011509470.1 | 892 | Silent Mutation | TAC,TAT | Y,Y 174 | XP_011507772.1 | |
XM_011509471.2 | 892 | Intron | XP_011507773.1 | |||
XM_017001123.1 | 892 | Silent Mutation | TAC,TAT | Y,Y 194 | XP_016856612.1 | |
XM_017001124.1 | 892 | Intron | XP_016856613.1 |
LOC730159 - uncharacterized LOC730159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195190.1 | 892 | Intron | NP_001182119.1 | |||
XM_011509968.2 | 892 | Intron | XP_011508270.1 |
Set Membership: |
HapMap |