Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGTGCCCCCTAATGCAGATAC[A/G]GCTAGAGTAACCAAAAACTTAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD45 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD45 - ankyrin repeat domain 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198493.2 | Intron | NP_940895.1 | ||||
XM_011509466.2 | Intron | XP_011507768.1 | ||||
XM_011509467.1 | Intron | XP_011507769.1 | ||||
XM_011509468.1 | Intron | XP_011507770.1 | ||||
XM_011509470.1 | Intron | XP_011507772.1 | ||||
XM_011509471.2 | Intron | XP_011507773.1 | ||||
XM_017001123.1 | Intron | XP_016856612.1 | ||||
XM_017001124.1 | Intron | XP_016856613.1 |
LOC730159 - uncharacterized LOC730159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195190.1 | Intron | NP_001182119.1 | ||||
XM_011509968.2 | Intron | XP_011508270.1 |
Set Membership: |
HapMap |