Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAGTTTCACTTTCTCCTCTTTT[C/G]CAGCTTTAAGAGTAAGGTCTACTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608783 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMYD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SMYD3 - SET and MYND domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167740.1 | Intron | NP_001161212.1 | ||||
NM_022743.2 | Intron | NP_073580.1 | ||||
XM_011544253.1 | Intron | XP_011542555.1 | ||||
XM_011544254.2 | Intron | XP_011542556.1 | ||||
XM_011544257.1 | Intron | XP_011542559.1 | ||||
XM_011544258.2 | Intron | XP_011542560.1 | ||||
XM_011544259.2 | Intron | XP_011542561.1 | ||||
XM_011544260.1 | Intron | XP_011542562.1 | ||||
XM_011544261.2 | Intron | XP_011542563.1 | ||||
XM_017002094.1 | Intron | XP_016857583.1 | ||||
XM_017002095.1 | Intron | XP_016857584.1 |
Set Membership: |
HapMap |