Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTGAATACACAGTATATTTTG[C/T]TATCCCTTCAAGTTATTAAATACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612686 | ||||||||||||||||||||
Literature Links: |
LOC283278 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC283278 - pleckstrin homology domain-containing family A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017018657.1 | 4535 | Intron | XP_016874146.1 | |||
XM_017018658.1 | 4535 | Intron | XP_016874147.1 |
PLEKHA7 - pleckstrin homology domain containing A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175058.4 | 4535 | UTR 3 | NP_778228.3 | |||
XM_017017240.1 | 4535 | Intron | XP_016872729.1 | |||
XM_017017241.1 | 4535 | UTR 3 | XP_016872730.1 | |||
XM_017017242.1 | 4535 | UTR 3 | XP_016872731.1 | |||
XM_017017243.1 | 4535 | UTR 3 | XP_016872732.1 |