Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGGGAGAAATGTCCAAATCTA[A/G]AGAATTCTCTTGTTTGTTGTTGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LYG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYG1 - lysozyme g1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174898.2 | Intron | NP_777558.1 | ||||
XM_005263869.2 | Intron | XP_005263926.1 | ||||
XM_005263870.3 | Intron | XP_005263927.1 | ||||
XM_005263871.4 | Intron | XP_005263928.1 | ||||
XM_011510580.2 | Intron | XP_011508882.1 | ||||
XM_017003311.1 | Intron | XP_016858800.1 | ||||
XM_017003312.1 | Intron | XP_016858801.1 | ||||
XM_017003313.1 | Intron | XP_016858802.1 |