Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATAGGATATCCTGAATTATCAA[A/G]GTGGGCCCTTAAATGTGGAAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612564 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LYG1 - lysozyme g1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC9 - thioredoxin domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005783.3 | Intron | NP_005774.2 | ||||
XM_017003147.1 | Intron | XP_016858636.1 |
Set Membership: |
HapMap |