Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCCTACCGTACCCCACCCTAT[C/T]TGCTCTACCCTTAGACAATTGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609953 | ||||||||||||||||||||
Literature Links: |
RAVER2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAVER2 - ribonucleoprotein, PTB binding 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018211.3 | Intron | NP_060681.2 | ||||
XM_006710738.3 | Intron | XP_006710801.2 | ||||
XM_011541706.2 | Intron | XP_011540008.1 |