Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATCTAGAAGAAGAGGACACTAT[C/T]GAGAGAAGGAGAAGAATATTCTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605836 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UNC13B - unc-13 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006377.3 | Intron | NP_006368.3 | ||||
XM_011517681.2 | Intron | XP_011515983.1 | ||||
XM_011517682.2 | Intron | XP_011515984.1 | ||||
XM_011517683.2 | Intron | XP_011515985.1 | ||||
XM_011517684.2 | Intron | XP_011515986.1 | ||||
XM_011517685.2 | Intron | XP_011515987.1 | ||||
XM_011517686.2 | Intron | XP_011515988.1 | ||||
XM_017014191.1 | Intron | XP_016869680.1 | ||||
XM_017014192.1 | Intron | XP_016869681.1 |