Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATTTGAGGGTATAAACCCATGG[A/C]TGGGCTCAGCTTTAAAAAAAAAAGA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604684 | |||||||||||||||||||||||||||||
Literature Links: |
GABPB2 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GABPB2 - GA binding protein transcription factor beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323906.1 | Intron | NP_001310835.1 | ||||
NM_001323907.1 | Intron | NP_001310836.1 | ||||
NM_001323908.1 | Intron | NP_001310837.1 | ||||
NM_001323909.1 | Intron | NP_001310838.1 | ||||
NM_001323910.1 | Intron | NP_001310839.1 | ||||
NM_001323911.1 | Intron | NP_001310840.1 | ||||
NM_001323912.1 | Intron | NP_001310841.1 | ||||
NM_001323913.1 | Intron | NP_001310842.1 | ||||
NM_144618.2 | Intron | NP_653219.1 | ||||
XM_017000246.1 | Intron | XP_016855735.1 | ||||
XM_017000247.1 | Intron | XP_016855736.1 | ||||
XM_017000248.1 | Intron | XP_016855737.1 | ||||
XM_017000249.1 | Intron | XP_016855738.1 | ||||
XM_017000250.1 | Intron | XP_016855739.1 | ||||
XM_017000251.1 | Intron | XP_016855740.1 | ||||
XM_017000252.1 | Intron | XP_016855741.1 | ||||
XM_017000253.1 | Intron | XP_016855742.1 | ||||
XM_017000254.1 | Intron | XP_016855743.1 |
MLLT11 - myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |