Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCATTTACTAAGATGACACGCT[A/G]ATTCGCTGCTTGGTGGAAACTCCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf107 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf107 - chromosome 10 open reading frame 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173554.2 | Intron | NP_775825.1 | ||||
XM_005269598.3 | Intron | XP_005269655.1 | ||||
XM_005269599.4 | Intron | XP_005269656.1 | ||||
XM_005269600.4 | Intron | XP_005269657.1 | ||||
XM_006717690.3 | Intron | XP_006717753.1 | ||||
XM_006717692.3 | Intron | XP_006717755.1 |