Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAAAAGAAAAGATAATGACCAA[A/G]TGTTTCACTTTATGAAATTCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611286 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTCA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RTCA - RNA 3'-terminal phosphate cyclase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130841.1 | Intron | NP_001124313.1 | ||||
NM_003729.3 | Intron | NP_003720.1 | ||||
XM_005271298.4 | Intron | XP_005271355.1 | ||||
XM_005271299.4 | Intron | XP_005271356.1 | ||||
XM_017002669.1 | Intron | XP_016858158.1 |
RTCA-AS1 - RTCA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |