Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTGTTTTTAAGGAACTTAGATG[A/G]TAGTGTGGAGCCAGGTGTGGTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606228 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AGO1 - argonaute 1, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317122.1 | Intron | NP_001304051.1 | ||||
NM_001317123.1 | Intron | NP_001304052.1 | ||||
NM_012199.4 | Intron | NP_036331.1 | ||||
XM_011541236.2 | Intron | XP_011539538.1 | ||||
XM_017001016.1 | Intron | XP_016856505.1 | ||||
XM_017001017.1 | Intron | XP_016856506.1 | ||||
XM_017001018.1 | Intron | XP_016856507.1 | ||||
XM_017001019.1 | Intron | XP_016856508.1 |
Set Membership: |
HapMap |