Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTTCCTTTGCCAACACTGTGT[A/G]CTATTCCAGCGATGTATCTTTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCMTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCMTD1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286782.1 | Intron | NP_001273711.1 | ||||
NM_001286783.1 | Intron | NP_001273712.1 | ||||
NM_052937.3 | Intron | NP_443169.2 | ||||
XM_005251146.2 | Intron | XP_005251203.1 | ||||
XM_011517444.2 | Intron | XP_011515746.1 | ||||
XM_017013017.1 | Intron | XP_016868506.1 |
Set Membership: |
HapMap |