Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCGTGTTGGTTCTGGTGAAGG[A/G]GGACATTCCTGCTCTTGAAATTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRAS2 - related RAS viral (r-ras) oncogene homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102669.2 | 715 | Silent Mutation | NP_001096139.1 | |||
NM_001177314.1 | 715 | Intron | NP_001170785.1 | |||
NM_001177315.1 | 715 | Intron | NP_001170786.1 | |||
NM_012250.5 | 715 | Silent Mutation | NP_036382.2 | |||
XM_017017363.1 | 715 | Intron | XP_016872852.1 |