Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGCCAACGGGGAGAAGCCCTTT[C/T]TGCTGCGGGCATTGCAGATCGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605881 | ||||||||||||||||||||
Literature Links: |
SLC35C1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC35C1 - solute carrier family 35 member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145265.1 | 815 | Silent Mutation | CTG,TTG | L,L 22 | NP_001138737.1 | |
NM_001145266.1 | 815 | Silent Mutation | CTG,TTG | L,L 22 | NP_001138738.1 | |
NM_018389.4 | 815 | Silent Mutation | CTG,TTG | L,L 35 | NP_060859.4 | |
XM_011520202.2 | 815 | Intron | XP_011518504.1 | |||
XM_011520203.2 | 815 | Silent Mutation | CTG,TTG | L,L 35 | XP_011518505.1 |