Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGAACTCAAGCAATCTGGCTTC[A/G]CAGTTCACATGCCCGACCACTGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605881 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35C1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC35C1 - solute carrier family 35 member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145265.1 | Intron | NP_001138737.1 | ||||
NM_001145266.1 | Intron | NP_001138738.1 | ||||
NM_018389.4 | Intron | NP_060859.4 | ||||
XM_011520202.2 | Intron | XP_011518504.1 | ||||
XM_011520203.2 | Intron | XP_011518505.1 |
Set Membership: |
HapMap |