Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGTCCAGCAGAGCTGTTATACA[C/T]GTGGTTAACACAATTACTGACTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603640 MIM: 609383 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADAM19 - ADAM metallopeptidase domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033274.4 | 5382 | UTR 3 | NP_150377.1 | |||
XM_005266003.2 | 5382 | Intron | XP_005266060.1 | |||
XM_011534682.2 | 5382 | UTR 3 | XP_011532984.1 | |||
XM_017010009.1 | 5382 | UTR 3 | XP_016865498.1 |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |