Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCACTGGAAAATGTCATGGCC[A/G]TAAGTATCTGCAGTCAGAAATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616467 MIM: 608071 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPCD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPCD - deleted in primary ciliary dyskinesia homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXW4 - F-box and WD repeat domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323541.1 | Intron | NP_001310470.1 | ||||
NM_022039.3 | Intron | NP_071322.1 | ||||
XM_005270053.2 | Intron | XP_005270110.2 | ||||
XM_017016570.1 | Intron | XP_016872059.1 |