Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTTTTAAAACTAGTTTAAAAG[A/G]TACATTACCCAAGAGTTGGGCCGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615536 MIM: 123660 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf80 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2orf80 - chromosome 2 open reading frame 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099334.2 | Intron | NP_001092804.1 | ||||
XM_017004074.1 | Intron | XP_016859563.1 | ||||
XM_017004075.1 | Intron | XP_016859564.1 | ||||
XM_017004076.1 | Intron | XP_016859565.1 | ||||
XM_017004077.1 | Intron | XP_016859566.1 | ||||
XM_017004078.1 | Intron | XP_016859567.1 |
CRYGA - crystallin gamma A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |