Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGATGACTTGCTCTTCTCATAG[A/C]CCATGGAGGCAAAGCTTAGGCTTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KIAA1551 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1551 - KIAA1551 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018169.3 | Intron | NP_060639.3 | ||||
XM_005253405.2 | Intron | XP_005253462.1 | ||||
XM_006719099.2 | Intron | XP_006719162.1 | ||||
XM_011520722.1 | Intron | XP_011519024.1 | ||||
XM_017019550.1 | Intron | XP_016875039.1 |