Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTAGATCTTCCACAGAGTGGA[A/C]TCTAAGGTGACTTAATTAATTGCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609531 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGRP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RASGRP3 - RAS guanyl releasing protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139488.1 | Intron | NP_001132960.1 | ||||
NM_015376.2 | Intron | NP_056191.1 | ||||
NM_170672.2 | Intron | NP_733772.1 | ||||
XM_005264246.3 | Intron | XP_005264303.1 | ||||
XM_011532746.2 | Intron | XP_011531048.1 | ||||
XM_011532747.2 | Intron | XP_011531049.1 | ||||
XM_011532748.2 | Intron | XP_011531050.2 | ||||
XM_017003759.1 | Intron | XP_016859248.1 | ||||
XM_017003760.1 | Intron | XP_016859249.1 | ||||
XM_017003761.1 | Intron | XP_016859250.1 | ||||
XM_017003762.1 | Intron | XP_016859251.1 | ||||
XM_017003763.1 | Intron | XP_016859252.1 |
Set Membership: |
HapMap |