Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCAGTAAAAGGAGGTGGGAAAC[C/G]TTGCTTTGAGGACAAATGTTGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613446 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP120 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP120 - centrosomal protein 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166226.1 | 4394 | UTR 3 | NP_001159698.1 | |||
NM_153223.3 | 4394 | UTR 3 | NP_694955.2 | |||
XM_005271901.4 | 4394 | UTR 3 | XP_005271958.1 | |||
XM_011543185.2 | 4394 | UTR 3 | XP_011541487.1 | |||
XM_011543186.2 | 4394 | UTR 3 | XP_011541488.1 | |||
XM_017009085.1 | 4394 | UTR 3 | XP_016864574.1 |