Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACCCCACCTCTCACAGCCTCTG[A/G]CCACCCTGCAGAGGGCCCAGCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600246 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELK4 - ELK4, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001973.3 | Intron | NP_001964.2 | ||||
NM_021795.2 | Intron | NP_068567.1 | ||||
XM_005244950.4 | Intron | XP_005245007.1 | ||||
XM_005244951.4 | Intron | XP_005245008.1 | ||||
XM_017000580.1 | Intron | XP_016856069.1 |
MFSD4A - major facilitator superfamily domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181644.4 | Intron | NP_857595.3 | ||||
XM_011509229.2 | Intron | XP_011507531.1 |
Set Membership: |
HapMap |