Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATACTCTATAAAACCAAACCC[A/C]ACCCTTCAATATTACACTAATGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612666 | ||||||||||||||||||||
Literature Links: |
DSTYK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSTYK - dual serine/threonine and tyrosine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015375.2 | 7752 | UTR 3 | NP_056190.1 | |||
NM_199462.2 | 7752 | UTR 3 | NP_955749.1 | |||
XM_011509392.2 | 7752 | UTR 3 | XP_011507694.1 | |||
XM_011509393.2 | 7752 | UTR 3 | XP_011507695.1 | |||
XM_011509394.2 | 7752 | UTR 3 | XP_011507696.1 |