Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATGTTGAACCAGTAAGATAAT[C/T]TAAGCATAAAGAAAAGGAAGCAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601275 | ||||||||||||||||||||
Literature Links: |
GPM6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPM6A - glycoprotein M6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261447.1 | 2858 | UTR 3 | NP_001248376.1 | |||
NM_001261448.1 | 2858 | UTR 3 | NP_001248377.1 | |||
NM_005277.4 | 2858 | UTR 3 | NP_005268.1 | |||
NM_201591.2 | 2858 | UTR 3 | NP_963885.1 | |||
NM_201592.2 | 2858 | UTR 3 | NP_963886.1 | |||
XM_006714183.1 | 2858 | UTR 3 | XP_006714246.1 | |||
XM_011531877.1 | 2858 | UTR 3 | XP_011530179.1 | |||
XM_017008036.1 | 2858 | UTR 3 | XP_016863525.1 |