Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACCGTGTACCTGCTGAGAGCAG[A/G]CACTTTCATTCAGTTTCTGTCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600804 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTNR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTNR1B - melatonin receptor 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005959.3 | Intron | NP_005950.1 | ||||
XM_011542839.2 | Intron | XP_011541141.1 | ||||
XM_017017777.1 | Intron | XP_016873266.1 |
Set Membership: |
HapMap |