Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTGTTCATCTAAAGATTTTAA[G/T]TTCATTTCTTTTAACAAAACTGAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612888 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC8B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC8B - leucine rich repeat containing 8 family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134476.1 | Intron | NP_001127948.1 | ||||
NM_015350.2 | Intron | NP_056165.1 | ||||
XM_005270701.4 | Intron | XP_005270758.1 | ||||
XM_005270702.4 | Intron | XP_005270759.1 | ||||
XM_005270703.4 | Intron | XP_005270760.1 | ||||
XM_011541143.2 | Intron | XP_011539445.1 | ||||
XM_011541144.2 | Intron | XP_011539446.1 | ||||
XM_011541145.2 | Intron | XP_011539447.1 | ||||
XM_011541146.2 | Intron | XP_011539448.1 | ||||
XM_011541148.2 | Intron | XP_011539450.1 | ||||
XM_017000883.1 | Intron | XP_016856372.1 | ||||
XM_017000884.1 | Intron | XP_016856373.1 | ||||
XM_017000885.1 | Intron | XP_016856374.1 | ||||
XM_017000886.1 | Intron | XP_016856375.1 |
Set Membership: |
HapMap |