Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAAACTTTACAAGATAAGGCTC[A/G]GAAGTCAGAAATCTTCAGGTTTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF749 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF749 - zinc finger protein 749 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023561.3 | Intron | NP_001018855.2 | ||||
NM_001321952.1 | Intron | NP_001308881.1 | ||||
NM_001321953.1 | Intron | NP_001308882.1 | ||||
NM_001321954.1 | Intron | NP_001308883.1 | ||||
XM_011526956.2 | Intron | XP_011525258.1 | ||||
XM_017026802.1 | Intron | XP_016882291.1 |
Set Membership: |
HapMap |