Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGTACACATGAGGGGAATGTA[C/T]ATTCTGTTGTTGTTGGGTAGAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601856 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF211 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF211 - zinc finger protein 211 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265597.2 | Intron | NP_001252526.1 | ||||
NM_001265598.2 | Intron | NP_001252527.1 | ||||
NM_001265599.2 | Intron | NP_001252528.1 | ||||
NM_001265600.2 | Intron | NP_001252529.1 | ||||
NM_001322306.1 | Intron | NP_001309235.1 | ||||
NM_006385.4 | Intron | NP_006376.2 | ||||
NM_198855.3 | Intron | NP_942152.1 |
Set Membership: |
HapMap |