Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTCAAAGGGGAACGTCAGGCA[A/G]AATGTCCCAGGGCTGCTGACTCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615847 MIM: 604259 | ||||||||||||||||||||
Literature Links: |
CEP83 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP83 - centrosomal protein 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042399.1 | Intron | NP_001035858.1 | ||||
NM_016122.2 | Intron | NP_057206.2 | ||||
XM_006719437.2 | Intron | XP_006719500.1 | ||||
XM_011538424.1 | Intron | XP_011536726.1 | ||||
XM_011538427.2 | Intron | XP_011536729.1 | ||||
XM_011538429.2 | Intron | XP_011536731.1 | ||||
XM_017019385.1 | Intron | XP_016874874.1 | ||||
XM_017019386.1 | Intron | XP_016874875.1 | ||||
XM_017019387.1 | Intron | XP_016874876.1 | ||||
XM_017019388.1 | Intron | XP_016874877.1 | ||||
XM_017019389.1 | Intron | XP_016874878.1 |
PLXNC1 - plexin C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005761.2 | Intron | NP_005752.1 | ||||
XM_006719186.3 | Intron | XP_006719249.1 | ||||
XM_011537730.2 | Intron | XP_011536032.1 | ||||
XM_011537731.2 | Intron | XP_011536033.1 | ||||
XM_017018671.1 | Intron | XP_016874160.1 | ||||
XM_017018672.1 | Intron | XP_016874161.1 |