Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613899 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCC - Fanconi anemia complementation group C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000136.2 | Intron | NP_000127.2 | ||||
NM_001243743.1 | Intron | NP_001230672.1 | ||||
NM_001243744.1 | Intron | NP_001230673.1 | ||||
XM_005251802.3 | Intron | XP_005251859.1 | ||||
XM_006717001.2 | Intron | XP_006717064.1 | ||||
XM_006717002.3 | Intron | XP_006717065.1 | ||||
XM_006717004.3 | Intron | XP_006717067.1 | ||||
XM_011518365.2 | Intron | XP_011516667.1 | ||||
XM_011518366.2 | Intron | XP_011516668.1 | ||||
XM_011518367.2 | Intron | XP_011516669.1 | ||||
XM_017014452.1 | Intron | XP_016869941.1 | ||||
XM_017014453.1 | Intron | XP_016869942.1 | ||||
XM_017014454.1 | Intron | XP_016869943.1 |