Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTATAGAAGTTGAAGTTTGACA[A/G]TTCCTCTGTCGTGCTTCATACTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616365 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEPDC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DEPDC4 - DEP domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319310.1 | Intron | NP_001306239.1 | ||||
NM_001319311.1 | Intron | NP_001306240.1 | ||||
NM_152317.3 | Intron | NP_689530.1 | ||||
XM_011537876.2 | Intron | XP_011536178.1 | ||||
XM_017018779.1 | Intron | XP_016874268.1 | ||||
XM_017018780.1 | Intron | XP_016874269.1 | ||||
XM_017018781.1 | Intron | XP_016874270.1 | ||||
XM_017018782.1 | Intron | XP_016874271.1 | ||||
XM_017018783.1 | Intron | XP_016874272.1 | ||||
XM_017018784.1 | Intron | XP_016874273.1 | ||||
XM_017018785.1 | Intron | XP_016874274.1 |
SCYL2 - SCY1 like pseudokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317784.1 | Intron | NP_001304713.1 | ||||
NM_017988.5 | Intron | NP_060458.3 | ||||
XM_017019618.1 | Intron | XP_016875107.1 | ||||
XM_017019619.1 | Intron | XP_016875108.1 | ||||
XM_017019620.1 | Intron | XP_016875109.1 |
Set Membership: |
HapMap |