Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGCCTTTTAAAGCATTTGATA[C/T]CTTCAAAGAAAAAATTCTGAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC17A8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC17A8 - solute carrier family 17 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145288.1 | 336 | Missense Mutation | ACC,ATC | T,I 8 | NP_001138760.1 | |
NM_139319.2 | 336 | Missense Mutation | ACC,ATC | T,I 8 | NP_647480.1 |