Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGATAAACCAAAATCTCAAGTAC[C/T]TGTTTTACAAAAAATTAAGTCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSB - arylsulfatase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000046.3 | 4509 | UTR 3 | NP_000037.2 | |||
NM_198709.2 | 4509 | Intron | NP_942002.1 | |||
XM_011543390.1 | 4509 | UTR 3 | XP_011541692.1 | |||
XM_011543391.2 | 4509 | Intron | XP_011541693.1 | |||
XM_011543392.2 | 4509 | Intron | XP_011541694.1 | |||
XM_011543393.1 | 4509 | Intron | XP_011541695.1 | |||
XM_017009471.1 | 4509 | Intron | XP_016864960.1 |