Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCCGCGGCACCATGGCACCTG[A/C]TGGACGCCCGGGGGCCAAGAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605932 MIM: 605849 | ||||||||||||||||||||
Literature Links: |
BHMT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BHMT2 - betaine--homocysteine S-methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178005.1 | 70 | Missense Mutation | GAT,GCT | D,A 4 | NP_001171476.1 | |
NM_017614.4 | 70 | Missense Mutation | GAT,GCT | D,A 4 | NP_060084.2 |
DMGDH - dimethylglycine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013391.3 | 70 | Intron | NP_037523.2 | |||
XM_006714597.2 | 70 | Intron | XP_006714660.1 | |||
XM_011543354.2 | 70 | Intron | XP_011541656.1 | |||
XM_011543355.2 | 70 | Intron | XP_011541657.1 |