Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAGGGACTAGATCTCAGAACT[A/C]ATTTCCTATCCTCACAGGTATTCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607730 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDIT4L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DDIT4L - DNA damage inducible transcript 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145244.3 | 1653 | UTR 3 | NP_660287.1 |
LOC101929353 - uncharacterized LOC101929353 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |