Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGAAGGAAAAACCAGAGCCCCC[A/G]GAACCGGAGCTGGGGACAGGACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605698 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378052 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105378052 - uncharacterized LOC105378052 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ULBP2 - UL16 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025217.3 | Intron | NP_079493.1 | ||||
XM_017011321.1 | Intron | XP_016866810.1 |
Set Membership: |
HapMap |