Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGGGCTGTGCCTGAGGACATGG[C/T]GGGTGGTGCTGAAATGGAAGCACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609244 | ||||||||||||||||||||
Literature Links: |
RAET1E-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAET1E-AS1 - RAET1E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAET1G - retinoic acid early transcript 1G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001788.3 | 782 | Missense Mutation | ACC,GCC | T,A 214 | NP_001001788.2 | |
XM_011535800.2 | 782 | Missense Mutation | ACC,GCC | T,A 214 | XP_011534102.1 | |
XM_017010831.1 | 782 | Missense Mutation | ACC,GCC | T,A 214 | XP_016866320.1 |