Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGGCTTTCCTTGTACATCTG[C/T]ATCACCTGTGGGGAGACACCAGCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603673 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTBD19 - BTB domain containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCH2 - patched 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166292.1 | 3375 | Missense Mutation | ATA,ATG | I,M 1121 | NP_001159764.1 | |
NM_003738.4 | 3375 | Missense Mutation | ATA,ATG | I,M 1121 | NP_003729.3 |