Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTLL4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTLL4 - tubulin tyrosine ligase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014640.4 | Intron | NP_055455.3 | ||||
XM_006712875.2 | Intron | XP_006712938.1 | ||||
XM_017005387.1 | Intron | XP_016860876.1 | ||||
XM_017005388.1 | Intron | XP_016860877.1 | ||||
XM_017005389.1 | Intron | XP_016860878.1 | ||||
XM_017005390.1 | Intron | XP_016860879.1 | ||||
XM_017005391.1 | Intron | XP_016860880.1 | ||||
XM_017005392.1 | Intron | XP_016860881.1 |
Set Membership: |
HapMap |