Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGACAGGTAGCTCCAGCTAAA[A/C]ATAAAAGTCTGGATGTAAGACTAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147522 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPKB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITPKB - inositol-trisphosphate 3-kinase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002221.3 | Intron | NP_002212.3 | ||||
XM_005273120.2 | Intron | XP_005273177.1 | ||||
XM_017001211.1 | Intron | XP_016856700.1 |
ITPKB-IT1 - ITPKB intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |