Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGCCTGTGGTCTCCTTCTGGTT[G/T]GGCAGGCGCCTCGTGGTTAGTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
CYP20A1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP20A1 - cytochrome P450 family 20 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177538.2 | 841 | Missense Mutation | TTG,TTT | L,F 73 | NP_803882.1 | |
XM_005246698.3 | 841 | Intron | XP_005246755.1 | |||
XM_005246699.3 | 841 | Intron | XP_005246756.1 | |||
XM_005246701.3 | 841 | Intron | XP_005246758.1 | |||
XM_006712644.3 | 841 | Missense Mutation | TTG,TTT | L,F 55 | XP_006712707.1 | |
XM_006712645.3 | 841 | Missense Mutation | TTG,TTT | L,F 73 | XP_006712708.1 | |
XM_017004555.1 | 841 | Intron | XP_016860044.1 |
Set Membership: |
DME Validated Inventoried |