Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGTGCAGGTTTGTTACATGGG[A/C]ATATTGTGTGGTACTGTGGTTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TET1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TET1 - tet methylcytosine dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030625.2 | Intron | NP_085128.2 | ||||
XM_011540204.2 | Intron | XP_011538506.1 | ||||
XM_011540205.2 | Intron | XP_011538507.1 | ||||
XM_011540206.2 | Intron | XP_011538508.1 | ||||
XM_011540207.2 | Intron | XP_011538509.1 | ||||
XM_017016686.1 | Intron | XP_016872175.1 | ||||
XM_017016687.1 | Intron | XP_016872176.1 | ||||
XM_017016688.1 | Intron | XP_016872177.1 | ||||
XM_017016689.1 | Intron | XP_016872178.1 |