Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCCATTTCGTTTTCATTGGCC[C/T]TCTTAGAAGATGATTAAGAAAAAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610373 MIM: 609397 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX50 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDX50 - DEAD-box helicase 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024045.1 | Intron | NP_076950.1 | ||||
XM_005270148.2 | Intron | XP_005270205.1 | ||||
XM_011540144.2 | Intron | XP_011538446.1 | ||||
XM_017016626.1 | Intron | XP_016872115.1 | ||||
XM_017016627.1 | Intron | XP_016872116.1 | ||||
XM_017016628.1 | Intron | XP_016872117.1 | ||||
XM_017016629.1 | Intron | XP_016872118.1 | ||||
XM_017016630.1 | Intron | XP_016872119.1 |
STOX1 - storkhead box 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |