Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCACTGAAGAGGTTGAGACAGTA[A/G]AGGTGGGTTGAAGCACTTTTAATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615335 MIM: 614478 | ||||||||||||||||||||
Literature Links: |
CERS5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CERS5 - ceramide synthase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281731.1 | 2166 | UTR 3 | NP_001268660.1 | |||
NM_147190.3 | 2166 | UTR 3 | NP_671723.1 | |||
XM_005269220.2 | 2166 | Intron | XP_005269277.1 | |||
XM_006719687.3 | 2166 | Intron | XP_006719750.1 | |||
XM_011538975.2 | 2166 | Intron | XP_011537277.1 | |||
XM_017020202.1 | 2166 | Intron | XP_016875691.1 | |||
XM_017020203.1 | 2166 | Intron | XP_016875692.1 | |||
XM_017020204.1 | 2166 | Intron | XP_016875693.1 | |||
XM_017020205.1 | 2166 | Intron | XP_016875694.1 |
COX14 - COX14, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |