Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTAAGGATATGACTGTAGCTA[G/T]ACCCTCTGGAAGTTCTCTTTATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609931 MIM: 609930 MIM: 601734 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987175 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC107987175 - uncharacterized LOC107987175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL6 - myosin light chain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021019.4 | Intron | NP_066299.2 | ||||
NM_079423.3 | Intron | NP_524147.2 |
MYL6B - myosin light chain 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCC2 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |