Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAAAATTTTGTTTTTGTTTTTG[G/T]TTGCATCCAAAACTGAAAGAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601545 | ||||||||||||||||||||
Literature Links: |
PAFAH1B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAFAH1B1 - platelet activating factor acetylhydrolase 1b regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000430.3 | 56 | Intron | NP_000421.1 | |||
XM_011523901.1 | 56 | Intron | XP_011522203.1 | |||
XM_011523902.2 | 56 | Intron | XP_011522204.1 | |||
XM_011523903.2 | 56 | Intron | XP_011522205.1 | |||
XM_017024699.1 | 56 | Intron | XP_016880188.1 | |||
XM_017024700.1 | 56 | Intron | XP_016880189.1 | |||
XM_017024701.1 | 56 | Intron | XP_016880190.1 | |||
XM_017024702.1 | 56 | Intron | XP_016880191.1 | |||
XM_017024703.1 | 56 | UTR 5 | XP_016880192.1 |