Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAATAATCAAAACTTGGGGGGG[A/G]AAACACCAAGTGACTGACATTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186360 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANXA7 - annexin A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001156.4 | Intron | NP_001147.1 | ||||
NM_001320879.1 | Intron | NP_001307808.1 | ||||
NM_001320880.1 | Intron | NP_001307809.1 | ||||
NM_004034.3 | Intron | NP_004025.1 | ||||
XM_017016162.1 | Intron | XP_016871651.1 | ||||
XM_017016163.1 | Intron | XP_016871652.1 |
Set Membership: |
HapMap |