Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTTCTTTTAAGATGTATCATTA[A/G]ATTGTTCATGTGAAGTTTTTTCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM149B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM149B1 - family with sequence similarity 149 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173348.1 | Intron | NP_775483.1 | ||||
XM_005269744.2 | Intron | XP_005269801.1 | ||||
XM_005269745.2 | Intron | XP_005269802.1 | ||||
XM_005269747.3 | Intron | XP_005269804.1 | ||||
XM_011539737.2 | Intron | XP_011538039.1 | ||||
XM_011539740.2 | Intron | XP_011538042.1 | ||||
XM_017016164.1 | Intron | XP_016871653.1 | ||||
XM_017016165.1 | Intron | XP_016871654.1 | ||||
XM_017016166.1 | Intron | XP_016871655.1 | ||||
XM_017016167.1 | Intron | XP_016871656.1 |